Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il5ra
Entrez Gene ID
16192
Gene Name
interleukin 5 receptor, alpha
Gene Aliases
CD125, CDw125, Il5r
Chromosome Location
Chr.6: 106710375 - 106749037 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC125374.1 - 10 - 11 1147 76
D90205.1 - 11 - 12 1381 76

View Full Product Details