Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kpnb1
Entrez Gene ID
16211
Gene Name
karyopherin (importin) beta 1
Gene Aliases
AA409963, IPOB, Impnb
Chromosome Location
Chr.11: 97159710 - 97187892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008379.3 NP_032405.3 3 - 4 588 68
GenBank mRNA AK077709.1 - 3 - 4 578 68
AK147175.1 - 3 - 4 607 68
AK169171.1 - 3 - 4 558 68
BC052438.1 - 3 - 4 343 68
BC052711.1 - 3 - 4 376 68
BC055115.1 - 3 - 4 573 68
D45836.1 - 3 - 4 479 68
D67015.1 - 3 - 4 316 68

View Full Product Details