The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008398.2 | NP_032424.2 | 1 - 2 | 385 | 100 |
GenBank mRNA | AK157612.1 | - | 1 - 2 | 380 | 100 |
BQ933931.1 | - | 1 - 2 | 151 | 100 | |
CN669314.1 | - | 1 - 2 | 367 | 100 | |
L23423.1 | - | 1 - 2 | 383 | 100 |