Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxyd3
Entrez Gene ID
17178
Gene Name
FXYD domain-containing ion transport regulator 3
Gene Aliases
Mat8
Chromosome Location
Chr.7: 31070535 - 31076697 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008557.2 NP_032583.1 3 - 4 159 76
GenBank mRNA AA959274.1 - 3 - 4 116 76
AK145206.1 - - 130 76
AK166426.1 - 3 - 4 132 76
BC002039.1 - 3 - 4 123 76
BC051033.1 - 3 - 4 117 76
BC056223.1 - 3 - 4 127 76
BG067532.2 - 5 - 6 76
X93038.1 - 3 - 4 155 76

View Full Product Details