The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008561.3 | NP_032587.1 | 1 - 1 | 1018 | 134 |
GenBank mRNA | AK038677.1 | - | 1 - 1 | 1019 | 134 |
BC103669.1 | - | 1 - 1 | 1007 | 134 | |
BC104730.1 | - | 1 - 1 | 1007 | 134 | |
BC105667.1 | - | 1 - 1 | 1007 | 134 | |
BC105668.1 | - | 1 - 1 | 1007 | 134 |