Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mertk
Entrez Gene ID
17289
Gene Name
c-mer proto-oncogene tyrosine kinase
Gene Aliases
Eyk, Mer, Nyk, nmf12
Chromosome Location
Chr.2: 128698997 - 128803162 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008587.1 NP_032613.1 7 - 8 1171 119
GenBank mRNA U21301.1 - 7 - 8 1171 119

View Full Product Details