Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Met
Entrez Gene ID
17295
Gene Name
met proto-oncogene
Gene Aliases
AI838057, HGF, HGFR, Par4, c-Met
Chromosome Location
Chr.6: 17463349 - 17573980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008591.2 NP_032617.2 21 - 22 4287 87
GenBank mRNA BC117969.1 - 19 - 20 3931 87
BC117970.1 - 19 - 20 3927 87
CX563675.1 - 2 - 3 245 87
Y00671.1 - 19 - 20 3932 87

View Full Product Details