The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163457.1 | NP_001156929.1 | 14 - 15 | 2157 | 116 |
NM_008642.2 | NP_032668.2 | 12 - 13 | 1847 | 116 | |
GenBank mRNA | AK146553.1 | - | 12 - 13 | 1848 | 116 |
BC012686.1 | - | 12 - 13 | 1807 | 116 | |
EU553486.1 | - | 12 - 13 | 1815 | 116 | |
L47970.1 | - | 12 - 13 | 1794 | 116 |