The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159520.1 | NP_001152992.1 | 7 - 8 | 1559 | 59 |
NM_008781.4 | NP_032807.3 | 7 - 8 | 1559 | 59 | |
GenBank mRNA | AK012493.1 | - | 7 - 8 | 1261 | 59 |
AK014337.1 | - | 7 - 8 | 1261 | 59 | |
AK044985.1 | - | 7 - 8 | 1264 | 59 | |
AK045018.1 | - | 7 - 8 | 1560 | 59 | |
AK144973.1 | - | 7 - 8 | 1264 | 59 | |
AK147919.1 | - | 7 - 8 | 1264 | 59 | |
AK148354.1 | - | 7 - 8 | 1540 | 59 | |
AK157760.1 | - | 7 - 8 | 1261 | 59 | |
AK191274.1 | - | 1 - 2 | 229 | 59 | |
AK214220.1 | - | 1 - 2 | 175 | 59 | |
BC048699.1 | - | 7 - 8 | 1230 | 59 | |
X59358.1 | - | 7 - 8 | 1469 | 59 |