The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129803.1 | NP_001123275.1 | 2 - 3 | 192 | 71 |
NM_001129804.1 | NP_001123276.1 | 2 - 3 | 164 | 71 | |
NM_008790.2 | NP_032816.1 | 2 - 3 | 189 | 71 | |
GenBank mRNA | AK003002.1 | - | 2 - 3 | 135 | 71 |
AK160398.1 | - | 2 - 3 | 153 | 71 | |
BC028982.1 | - | 2 - 3 | 172 | 71 | |
BU531373.1 | - | 2 - 3 | 190 | 71 | |
CB522989.1 | - | 2 - 3 | 164 | 71 | |
EU164759.1 | - | 2 - 3 | 192 | 71 | |
M21532.1 | - | 2 - 3 | 159 | 71 |