Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pik3cd
Entrez Gene ID
18707
Gene Name
phosphatidylinositol 3-kinase catalytic delta polypeptide
Gene Aliases
2410099E07Rik, 2610208K16Rik, AW545373, p110delta
Chromosome Location
Chr.4: 149649168 - 149702566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029837.2 NP_001025008.2 10 - 11 1614 71
NM_001164049.1 NP_001157521.1 10 - 11 1641 71
NM_001164050.1 NP_001157522.1 10 - 11 1641 71
NM_001164051.1 NP_001157523.1 10 - 11 1665 71
NM_001164052.1 NP_001157524.1 8 - 9 1368 71
NM_008840.3 NP_032866.2 11 - 12 1816 71
GenBank mRNA AK040867.1 - 10 - 11 1576 71
AK149935.1 - 11 - 12 1715 71
AK150649.1 - 10 - 11 1542 71
AK151482.1 - 10 - 11 1542 71
AK152661.1 - 10 - 11 1542 71
AK153304.1 - 10 - 11 1564 71
AK155835.1 - 10 - 11 1527 71
AK171025.1 - 10 - 11 1624 71
AK172222.1 - 10 - 11 1518 71
BC035203.1 - 10 - 11 1631 71
CT010349.1 - 8 - 9 1336 71
U86587.1 - 8 - 9 1336 71

View Full Product Details