Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pik3cd
Entrez Gene ID
18707
Gene Name
phosphatidylinositol 3-kinase catalytic delta polypeptide
Gene Aliases
2410099E07Rik, 2610208K16Rik, AW545373, p110delta
Chromosome Location
Chr.4: 149649168 - 149702566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029837.2 NP_001025008.2 14 - 15 2098 67
NM_001164049.1 NP_001157521.1 14 - 15 2125 67
NM_001164050.1 NP_001157522.1 14 - 15 2116 67
NM_001164051.1 NP_001157523.1 14 - 15 2146 67
NM_001164052.1 NP_001157524.1 12 - 13 1840 67
NM_008840.3 NP_032866.2 15 - 16 2291 67
GenBank mRNA AK040867.1 - 14 - 15 2060 67
AK149935.1 - 15 - 16 2190 67
AK153304.1 - 14 - 15 2039 67
AK155835.1 - 14 - 15 2003 67
AK171025.1 - 14 - 15 2105 67
AK172222.1 - 14 - 15 2002 67
BC035203.1 - 14 - 15 2106 67
CT010349.1 - 12 - 13 1811 67
U86587.1 - 12 - 13 1808 67

View Full Product Details