The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029837.2 | NP_001025008.2 | 16 - 17 | 2341 | 61 |
NM_001164049.1 | NP_001157521.1 | 16 - 17 | 2368 | 61 | |
NM_001164050.1 | NP_001157522.1 | 16 - 17 | 2359 | 61 | |
NM_001164051.1 | NP_001157523.1 | 16 - 17 | 2389 | 61 | |
NM_001164052.1 | NP_001157524.1 | 14 - 15 | 2083 | 61 | |
NM_008840.3 | NP_032866.2 | 17 - 18 | 2534 | 61 | |
GenBank mRNA | AK040867.1 | - | 16 - 17 | 2303 | 61 |
AK149935.1 | - | 17 - 18 | 2433 | 61 | |
AK153304.1 | - | 16 - 17 | 2282 | 61 | |
AK155835.1 | - | 16 - 17 | 2246 | 61 | |
AK171025.1 | - | 16 - 17 | 2348 | 61 | |
AK172222.1 | - | 16 - 17 | 2245 | 61 | |
BC035203.1 | - | 16 - 17 | 2349 | 61 | |
CT010349.1 | - | 14 - 15 | 2054 | 61 | |
U86587.1 | - | 14 - 15 | 2051 | 61 |