The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008858.3 | NP_032884.2 | 3 - 4 | 776 | 126 |
GenBank mRNA | Z34524.1 | - | 3 - 4 | 651 | 126 |