The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008926.4 | NP_032952.3 | 15 - 16 | 2108 | 99 |
GenBank mRNA | AK038739.1 | - | 15 - 16 | 2092 | 99 |
AK081422.1 | - | 15 - 16 | 2109 | 99 | |
BC113205.1 | - | 15 - 16 | 2019 | 99 | |
L12460.1 | - | 15 - 16 | 2380 | 99 |