Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptch1
Entrez Gene ID
19206
Gene Name
patched homolog 1
Gene Aliases
A230106A15Rik, Ptc, Ptc1, Ptch, mes
Chromosome Location
Chr.13: 63508328 - 63573616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008957.2 NP_032983.1 17 - 18 2846 69
GenBank mRNA AK147626.1 - 18 - 19 2993 69
U46155.1 - 17 - 18 2846 69

View Full Product Details