The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008957.2 | NP_032983.1 | 22 - 23 | 3768 | 135 |
GenBank mRNA | AK147626.1 | - | 23 - 24 | 3915 | 135 |
AK163736.1 | - | 2 - 3 | 337 | 135 | |
U46155.1 | - | 22 - 23 | 3768 | 135 |