The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166406.1 | NP_001159878.1 | 2 - 3 | 412 | 110 |
NM_001166407.1 | NP_001159879.1 | 2 - 3 | 410 | 110 | |
NM_009004.4 | NP_033030.1 | 2 - 3 | 306 | 110 | |
GenBank mRNA | AK051201.1 | - | 2 - 3 | 224 | 110 |
AK051995.1 | - | 2 - 3 | 261 | 110 | |
AK053280.1 | - | 2 - 3 | 224 | 110 | |
AK083412.1 | - | 2 - 3 | 266 | 110 | |
AK083697.1 | - | 2 - 3 | 226 | 110 | |
AK084732.1 | - | 2 - 3 | 248 | 110 | |
AK142764.1 | - | 2 - 3 | 236 | 110 | |
AK144844.1 | - | 2 - 3 | 224 | 110 | |
BC060608.1 | - | 2 - 3 | 412 | 110 | |
Y09632.1 | - | 2 - 3 | 461 | 110 |