The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080780.1 | NP_001074249.1 | 2 - 3 | 954 | 67 |
NM_009050.2 | NP_033076.2 | 2 - 3 | 954 | 67 | |
GenBank mRNA | AF209436.1 | - | 2 - 3 | 343 | 67 |
AK051633.1 | - | 2 - 3 | 533 | 67 | |
AK084977.1 | - | 2 - 3 | 603 | 67 | |
AY326397.1 | - | 2 - 3 | 343 | 67 | |
BC059012.1 | - | 2 - 3 | 928 | 67 | |
X67812.1 | - | 2 - 3 | 343 | 67 |