Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ralgds
Entrez Gene ID
19730
Gene Name
ral guanine nucleotide dissociation stimulator
Gene Aliases
Gnds, Rgds, mKIAA1308
Chromosome Location
Chr.2: 28513089 - 28553082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145834.1 NP_001139306.1 12 - 13 1946 91
NM_001145835.1 NP_001139307.1 12 - 13 1858 91
NM_001145836.1 NP_001139308.1 12 - 13 1822 91
NM_009058.2 NP_033084.2 12 - 13 1982 91
GenBank mRNA AK129328.1 - 12 - 13 1823 91
AK141959.1 - 12 - 13 1813 91
AK149644.1 - 7 - 8 717 91
AK159020.1 - 12 - 13 1815 91
AK159754.1 - 12 - 13 1828 91
BC027225.1 - 12 - 13 1640 91
L07924.1 - 12 - 13 1818 91

View Full Product Details