The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159731.1 | NP_001153203.1 | 2 - 3 | 374 | 65 |
NM_009107.3 | NP_033133.1 | 3 - 4 | 830 | 65 | |
GenBank mRNA | BC058401.1 | - | 3 - 4 | 812 | 65 |
BM947610.1 | - | 3 - 4 | 556 | 65 | |
G49237.1 | - | 3 - 4 | 562 | 65 | |
M84819.1 | - | 3 - 4 | 475 | 65 | |
S62948.1 | - | 3 - 4 | 674 | 65 | |
S62952.1 | - | 2 - 3 | 330 | 65 | |
X66225.1 | - | 3 - 4 | 562 | 65 |