Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrx2
Entrez Gene ID
20204
Gene Name
paired related homeobox 2
Gene Aliases
Prx2, S8
Chromosome Location
Chr.2: 30845064 - 30881251 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009116.2 NP_033142.2 2 - 3 497 56
GenBank mRNA AK019971.1 - - 464 56
BC137872.1 - 2 - 3 524 56
BC137874.1 - 2 - 3 524 56
CB193470.1 - 1 - 2 172 56
X14572.1 - - 78 56
X52875.1 - 2 - 3 584 56

View Full Product Details