The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009120.2 | NP_033146.1 | 1 - 2 | 92 | 118 |
GenBank mRNA | AK157843.1 | - | 1 - 2 | 104 | 118 |
AK159239.1 | - | 1 - 2 | 103 | 118 | |
AK168175.1 | - | 1 - 2 | 118 | ||
BC005549.1 | - | 1 - 2 | 93 | 118 |