The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243072.1 | NP_001230001.1 | 15 - 16 | 2034 | 75 |
NM_001243073.1 | NP_001230002.1 | 15 - 16 | 1874 | 75 | |
NM_009152.4 | NP_033178.2 | 14 - 15 | 2302 | 75 | |
GenBank mRNA | AK140817.1 | - | 15 - 16 | 1875 | 75 |
BC057588.1 | - | 14 - 15 | 1949 | 75 | |
BC090844.1 | - | 15 - 16 | 2034 | 75 | |
D85028.2 | - | 14 - 15 | 2304 | 75 | |
L40484.1 | - | 11 - 12 | 1335 | 75 | |
L41541.1 | - | 14 - 15 | 1653 | 75 | |
X85993.1 | - | 14 - 15 | 1763 | 75 |