The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009257.3 | NP_033283.1 | 5 - 6 | 643 | 126 |
GenBank mRNA | AK136532.1 | - | 5 - 6 | 649 | 126 |
AK145220.1 | - | 4 - 5 | 611 | 126 | |
BC005434.1 | - | 5 - 6 | 644 | 126 | |
U54705.1 | - | 6 - 7 | 750 | 126 |