Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stc1
Entrez Gene ID
20855
Gene Name
stanniocalcin 1
Gene Aliases
Stc
Chromosome Location
Chr.14: 69029289 - 69041401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009285.3 NP_033311.3 2 - 3 366 105
GenBank mRNA AF099098.1 - 2 - 3 418 105
AK134260.1 - 2 - 3 367 105
AK142220.1 - 2 - 3 265 105
AK149293.1 - 2 - 3 249 105
AK149351.1 - 2 - 3 249 105
AK161960.1 - 2 - 3 248 105
BC021425.1 - 2 - 3 387 105

View Full Product Details