Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk10
Entrez Gene ID
20868
Gene Name
serine/threonine kinase 10
Gene Aliases
Gek1, Lok, mKIAA4026
Chromosome Location
Chr.11: 32533266 - 32624595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009288.2 NP_033314.2 16 - 17 2660 72
GenBank mRNA AK148118.1 - 16 - 17 2600 72
AK220448.1 - 16 - 17 2582 72
BC069840.1 - 16 - 17 2733 72
BC096624.1 - 16 - 17 2745 72
BC128363.1 - 16 - 17 2546 72
D89728.1 - 16 - 17 2621 72

View Full Product Details