The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286628.1 | NP_001273557.1 | 8 - 9 | 1045 | 109 |
NM_009352.3 | NP_033378.1 | 9 - 10 | 1132 | 109 | |
GenBank mRNA | AB109563.1 | - | 9 - 10 | 1112 | 109 |
AK132022.1 | - | 8 - 9 | 1028 | 109 | |
BC034866.1 | - | 9 - 10 | 1120 | 109 | |
U65586.1 | - | 9 - 10 | 1112 | 109 |