The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009392.2 | NP_033418.1 | 1 - 2 | 546 | 62 |
GenBank mRNA | AK013072.1 | - | 1 - 2 | 546 | 62 |
BC138240.1 | - | 1 - 2 | 420 | 62 | |
BC138241.1 | - | 1 - 2 | 420 | 62 | |
M75953.1 | - | 1 - 2 | 464 | 62 |