Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tshb
Entrez Gene ID
22094
Gene Name
thyroid stimulating hormone, beta subunit
Gene Aliases
-
Chromosome Location
Chr.3: 102773624 - 102803490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009432.2 NP_033458.2 3 - 4 152 109
GenBank mRNA BC116829.1 - 2 - 3 73 109
BC116831.1 - 2 - 3 73 109
J00644.1 - 2 - 3 85 109
M54943.1 - 2 - 3 85 109

View Full Product Details