The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190974.1 | NP_001177903.1 | 6 - 7 | 1021 | 70 |
NM_001190975.1 | NP_001177904.1 | 6 - 7 | 1021 | 70 | |
NM_009465.4 | NP_033491.2 | 6 - 7 | 1021 | 70 | |
GenBank mRNA | AK033237.1 | - | 6 - 7 | 990 | 70 |
AK155567.1 | - | 7 - 8 | 907 | 70 | |
AK170010.1 | - | 6 - 7 | 995 | 70 | |
AK170897.1 | - | 7 - 8 | 918 | 70 | |
BC008551.1 | - | 1 - 2 | 111 | 70 | |
BC046618.1 | - | 6 - 7 | 858 | 70 | |
BC050914.1 | - | 6 - 7 | 829 | 70 | |
BC058230.1 | - | 7 - 8 | 909 | 70 | |
X63535.1 | - | 6 - 7 | 770 | 70 |