The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009504.4 | NP_033530.2 | 8 - 9 | 1028 | 94 |
GenBank mRNA | AK030339.1 | - | 8 - 9 | 1029 | 94 |
AK154647.1 | - | 8 - 9 | 1003 | 94 | |
AK156789.1 | - | 8 - 9 | 1044 | 94 | |
BC006716.1 | - | 8 - 9 | 994 | 94 | |
CT010294.1 | - | 6 - 7 | 888 | 94 | |
D31969.1 | - | 8 - 9 | 996 | 94 |