The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171739.1 | NP_001165210.1 | 1 - 2 | 571 | 81 |
NM_009736.3 | NP_033866.4 | 1 - 2 | 571 | 81 | |
GenBank mRNA | AF022223.1 | - | 1 - 2 | 512 | 81 |
AK009149.1 | - | 1 - 2 | 482 | 81 | |
AK154469.1 | - | 1 - 2 | 521 | 81 | |
AK171094.1 | - | 1 - 2 | 521 | 81 | |
BC003722.1 | - | 1 - 2 | 533 | 81 | |
BC069918.1 | - | 1 - 2 | 508 | 81 | |
BC093509.1 | - | 1 - 2 | 482 | 81 | |
CA321856.1 | - | 1 - 2 | 558 | 81 | |
CX225744.1 | - | 1 - 2 | 571 | 81 |