Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspo
Entrez Gene ID
12257
Gene Name
translocator protein
Gene Aliases
Bzrp, IBP, PBR, Tspo1
Chromosome Location
Chr.15: 83563573 - 83574203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009775.4 NP_033905.3 3 - 4 420 116
GenBank mRNA AK149743.1 - 3 - 4 395 116
AK150516.1 - 3 - 4 392 116
AK151421.1 - 3 - 4 394 116
AK151569.1 - 3 - 4 401 116
AK151824.1 - 3 - 4 401 116
AK152082.1 - 3 - 4 392 116
AK152827.1 - 3 - 4 392 116
AK153159.1 - 3 - 4 394 116
AK153273.1 - 3 - 4 404 116
AK156489.1 - 3 - 4 395 116
AK165450.1 - 3 - 4 394 116
AK165822.1 - 3 - 4 401 116
AK166125.1 - 3 - 4 401 116
AK172384.1 - 3 - 4 395 116
BC002055.1 - 3 - 4 382 116
D21207.1 - 3 - 4 384 116
L17306.1 - 2 - 3 349 116

View Full Product Details