The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009780.2 | NP_033910.2 | 14 - 15 | 1935 | 75 |
GenBank mRNA | AK128974.1 | - | 2 - 3 | 361 | 75 |
AK157954.1 | - | 14 - 15 | 1921 | 75 | |
AK158256.1 | - | 14 - 15 | 1921 | 75 | |
AK158475.1 | - | 14 - 15 | 1921 | 75 | |
AK171644.1 | - | 14 - 15 | 1921 | 75 | |
AK172637.1 | - | 14 - 15 | 1924 | 75 | |
BC039141.1 | - | 14 - 15 | 1898 | 75 | |
BC055297.1 | - | 14 - 15 | 1905 | 75 | |
BC067394.1 | - | 14 - 15 | 1905 | 75 | |
BC067409.1 | - | 14 - 15 | 1905 | 75 | |
BC150743.1 | - | 14 - 15 | 1885 | 75 | |
M11729.1 | - | 14 - 15 | 1920 | 75 | |
X05314.1 | - | 1 - 2 | 94 | 75 |