The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009787.2 | NP_033917.2 | 9 - 10 | 1720 | 69 |
GenBank mRNA | AK028292.1 | - | 9 - 10 | 1570 | 69 |
AK146288.1 | - | 9 - 10 | 1553 | 69 | |
AK148801.1 | - | 9 - 10 | 1642 | 69 | |
AK150566.1 | - | 9 - 10 | 1601 | 69 | |
AK150903.1 | - | 1 - 2 | 228 | 69 | |
AK161534.1 | - | 9 - 10 | 1558 | 69 | |
AK168447.1 | - | 9 - 10 | 1524 | 69 | |
AK169387.1 | - | 9 - 10 | 1565 | 69 | |
BC066857.1 | - | 8 - 9 | 1319 | 69 | |
BC141078.1 | - | 9 - 10 | 1616 | 69 | |
J05186.1 | - | 9 - 10 | 1603 | 69 | |
Y00884.1 | - | 8 - 9 | 1426 | 69 |