Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd6
Entrez Gene ID
12511
Gene Name
CD6 antigen
Gene Aliases
-
Chromosome Location
Chr.19: 10789339 - 10830059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037801.2 NP_001032890.1 2 - 3 319 62
NM_009852.3 NP_033982.3 2 - 3 319 62
GenBank mRNA AK030822.1 - 2 - 3 289 62
AK030823.1 - 2 - 3 320 62
AK088050.1 - 2 - 3 297 62
BC014294.1 - 2 - 3 325 62
U12434.1 - 2 - 3 117 62
U35370.1 - 2 - 3 235 62
U37543.1 - 2 - 3 276 62
U37544.1 - 2 - 3 276 62

View Full Product Details