The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009854.2 | NP_033984.1 | 3 - 4 | 635 | 97 |
GenBank mRNA | AK136444.1 | - | 3 - 4 | 589 | 97 |
AK154339.1 | - | 3 - 4 | 590 | 97 | |
D10329.1 | - | 3 - 4 | 575 | 97 |