The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009897.2 | NP_034027.1 | 4 - 5 | 881 | 89 |
GenBank mRNA | AK005076.1 | - | 4 - 5 | 880 | 89 |
AK131953.1 | - | 4 - 5 | 885 | 89 | |
BC025976.1 | - | 5 - 6 | 796 | 89 | |
Z13968.1 | - | 4 - 5 | 881 | 89 |