The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009897.2 | NP_034027.1 | 6 - 7 | 1088 | 84 |
GenBank mRNA | AK005076.1 | - | 6 - 7 | 1087 | 84 |
AK131953.1 | - | 6 - 7 | 1092 | 84 | |
BC025976.1 | - | 7 - 8 | 1003 | 84 | |
Z13968.1 | - | 6 - 7 | 1088 | 84 |