The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009971.1 | NP_034101.1 | 3 - 4 | 389 | 63 |
GenBank mRNA | BC120759.1 | - | 3 - 4 | 342 | 63 |
BC120761.1 | - | 3 - 4 | 342 | 63 | |
M13926.1 | - | 3 - 4 | 389 | 63 |