Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 4 - 5 738 104
GenBank mRNA AF012251.1 - 4 - 5 463 104
AJ001420.1 - 4 - 5 708 104
AK002927.1 - 4 - 5 746 104
AK076042.1 - 4 - 5 699 104
BC047143.1 - 4 - 5 728 104
BC057862.1 - 4 - 5 398 104

View Full Product Details