Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 6 - 7 888 93
GenBank mRNA AF012251.1 - 6 - 7 613 93
AJ001420.1 - 6 - 7 858 93
AK002927.1 - 6 - 7 896 93
AK076042.1 - 6 - 7 849 93
BC047143.1 - 6 - 7 878 93
BC057862.1 - 6 - 7 548 93

View Full Product Details