Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 7 - 8 992 78
GenBank mRNA AF012251.1 - 7 - 8 717 78
AJ001420.1 - 7 - 8 962 78
AK002927.1 - 7 - 8 1000 78
AK076042.1 - 7 - 8 953 78
BC047143.1 - 7 - 8 982 78
BC057862.1 - 7 - 8 652 78

View Full Product Details