Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgcr2
Entrez Gene ID
13356
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
9930034O06Rik, DGS-C, Dgsc, Idd, Lan, Sez12, mKIAA0163
Chromosome Location
Chr.16: 17840355 - 17894098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109750.1 NP_001103220.1 9 - 10 1651 95
NM_010048.3 NP_034178.2 9 - 10 1660 95
GenBank mRNA AK088131.1 - 8 - 9 1377 95
AK129070.1 - 10 - 11 1510 95
AK133880.1 - 6 - 7 949 95
AK141594.1 - 9 - 10 1626 95
AK145495.1 - 9 - 10 1577 95
AK153026.1 - 9 - 10 1447 95
AK161022.1 - - 2411 95
AK161359.1 - 9 - 10 1376 95
AK163872.1 - 9 - 10 1613 95
AK163903.1 - 9 - 10 1589 95
AK164263.1 - 2 - 3 360 95
AK192000.1 - 1 - 2 69 95
BC031506.1 - 2 - 3 370 95
BC060636.1 - 9 - 10 1601 95
BC062978.1 - 9 - 10 1555 95
D78641.1 - 9 - 10 1585 95
X95480.1 - 9 - 10 1526 95

View Full Product Details