The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010062.3 | NP_034192.1 | 1 - 2 | 250 | 71 |
GenBank mRNA | AK145192.1 | - | 1 - 2 | 251 | 71 |
BC058609.1 | - | 1 - 2 | 197 | 71 | |
BQ829838.1 | - | 1 - 2 | 236 | 71 |