Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dok2
Entrez Gene ID
13449
Gene Name
docking protein 2
Gene Aliases
DokR, Frip
Chromosome Location
Chr.14: 70764528 - 70778494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010071.2 NP_034201.1 2 - 3 409 85
GenBank mRNA AF030627.1 - 2 - 3 409 85
AF035117.1 - 2 - 3 405 85
AF059583.1 - 2 - 3 395 85
AK158246.1 - 2 - 3 373 85
AK159466.1 - 2 - 3 450 85
AK159472.1 - 2 - 3 487 85
BC004590.1 - 2 - 3 401 85

View Full Product Details