The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177937.1 | NP_001171408.1 | 5 - 6 | 916 | 62 |
NM_001177938.1 | NP_001171409.1 | 5 - 6 | 916 | 62 | |
NM_001177939.1 | NP_001171410.1 | 5 - 6 | 916 | 62 | |
NM_001177940.1 | NP_001171411.1 | 4 - 5 | 892 | 62 | |
NM_001177941.1 | NP_001171412.1 | 4 - 5 | 892 | 62 | |
NM_001177942.1 | NP_001171413.1 | 5 - 6 | 916 | 62 | |
NM_001177943.1 | NP_001171414.1 | 5 - 6 | 916 | 62 | |
NM_001177944.1 | NP_001171415.1 | 5 - 6 | 916 | 62 | |
NM_010099.2 | NP_034229.1 | 5 - 6 | 916 | 62 | |
GenBank mRNA | AF004435.1 | - | 5 - 6 | 882 | 62 |
AF016628.1 | - | 5 - 6 | 916 | 62 | |
AF016629.1 | - | 5 - 6 | 916 | 62 | |
AJ243657.1 | - | 5 - 6 | 903 | 62 | |
AJ243658.1 | - | 5 - 6 | 864 | 62 | |
AK137551.1 | - | 5 - 6 | 825 | 62 | |
BC125387.1 | - | 5 - 6 | 807 | 62 | |
BC144790.1 | - | 5 - 6 | 807 | 62 | |
BC144791.1 | - | 5 - 6 | 807 | 62 | |
DQ168876.1 | - | 5 - 6 | 916 | 62 | |
DQ168877.1 | - | 5 - 6 | 916 | 62 | |
DQ168878.1 | - | 4 - 5 | 892 | 62 | |
DQ168879.1 | - | 4 - 5 | 892 | 62 | |
DQ168880.1 | - | 5 - 6 | 916 | 62 | |
DQ168881.1 | - | 5 - 6 | 916 | 62 |