The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010134.3 | NP_034264.1 | 1 - 2 | 1491 | 84 |
GenBank mRNA | AK157766.1 | - | 1 - 2 | 1493 | 84 |
L12705.1 | - | 1 - 2 | 707 | 84 |