Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabra1
Entrez Gene ID
14394
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit alpha 1
Gene Aliases
Gabra-1
Chromosome Location
Chr.11: 42130939 - 42182951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010250.5 NP_034380.1 2 - 3 857 109
GenBank mRNA AK034095.1 - 2 - 3 858 109
AK039496.1 - 2 - 3 141 109
AK141596.1 - 2 - 3 717 109
BC132331.1 - 2 - 3 533 109
BC137733.1 - 2 - 3 536 109
M63436.1 - 2 - 3 530 109
M86566.1 - 2 - 3 225 109
X61430.1 - 1 - 2 113 109

View Full Product Details